p-Cymene, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ100
C121452-1L
C121452-25ML
C121452-4L
SIALC121452-1LEA
67.6
EUR
SIALC121452-1L
SIALC121452-25ML
SIALC121452-4L
p-Cymene, Sigma-Aldrich®
p-Cymene
Formula:
C₁₀H₁₄ Boiling Pt: 176 °C (1013 hPa) Melting Pt: –67.9 °C Density: 0.8573 g/cm³ (20 °C) Flash Pt: 47 °C |
MDL Number:
MFCD00008893 CAS Number: 99-87-6 EINECS: 202-796-7 UN: 2046 ADR: 3,III Merck Index: 13,02792 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...