4-Methylbenzophenone, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
M29959-10G M29959-50G
SIALM29959-10GEA 27.1 EUR
SIALM29959-10G SIALM29959-50G
4-Methylbenzophenone, Sigma-Aldrich®
4-Methylbenzophenone
Formula: C₁₄H₁₂O
Boiling Pt: 326 °C (1013 hPa)
Melting Pt: 53…57 °C
Density: 0.9926 g/cm³ (20 °C)
MDL Number: MFCD00008553
CAS Number: 134-84-9
EINECS: 205-159-1
Merck Index: 12,07470