4,4'-Bis(diethylamino)benzophenone ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
B22288.14
B22288.22
B22288.14EA
73.4
EUR
B22288.14
B22288.22
4,4'-Bis(diethylamino)benzophenone ≥98%
4,4'-Bis(diethylamino)benzophenone
Purity: 99%
CAS No.: 90-93-7
Molecular Formula: C21H28N2O
Formula Weight: 324.47
Flash Point: 151°(303°F)
Odour: Odourless
pH: 4-6
Melting Point: 93-95°
Beilstein Reference: 2148611
UN No.: 3077
MDL No.: MFCD00009044
Formula:
C₂₁H₂₈N₂O Boiling Pt: 420 °C (1013 hPa) Melting Pt: 93…95 °C Flash Pt: 151 °C (303 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009044 CAS Number: 90-93-7 EINECS: 202-025-4 UN: 3077 ADR: 9,III |