1,2,4-Trimethylbenzene, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Pseudocumene
T73601-25ML T73601-2L T73601-500ML
SIALT73601-25MLEA 34.2 EUR
SIALT73601-25ML SIALT73601-2L SIALT73601-500ML
1,2,4-Trimethylbenzene, Sigma-Aldrich®
1,2,4-Trimethylbenzene
Formula: C₉H₁₂
Boiling Pt: 169 °C (1013 hPa)
Melting Pt: –44 °C
Density: 0.88 g/cm³ (20 °C)
Flash Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00008527
CAS Number: 95-63-6
EINECS: 202-436-9
UN: 3295
ADR: 3,III
Merck Index: 12,08100