1,3,5-Tri-tert-butylbenzene, Sigma-Aldrich®
Supplier: Merck
223778-10G
223778-50G
SIAL223778-10GEA
306
EUR
SIAL223778-10G
SIAL223778-50G
1,3,5-Tri-tert-butylbenzene, Sigma-Aldrich®
1,3,5-Tri-tert-butylbenzene
Formula:
C₁₈H₃₀ Boiling Pt: 248 °C (1013 hPa) Melting Pt: 69…71 °C Density: 2,637 g/cm³ (22 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008831 CAS Number: 1460-02-2 EINECS: 215-952-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...