1,2,4,5-Tetramethylbenzene, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

T19607-100G T19607-500G
SIALT19607-100GEA 59 EUR
SIALT19607-100G SIALT19607-500G
1,2,4,5-Tetramethylbenzene, Sigma-Aldrich®
1,2,4,5-Tetramethylbenzene
Formula: C₁₀H₁₄
Boiling Pt: 196.8 °C (1013 hPa)
Melting Pt: 78…81 °C
Density: 1.036 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008528
CAS Number: 95-93-2
EINECS: 202-465-7
UN: 1325
ADR: 4.1,II
Merck Index: 13,03501

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR