Cyclohexen-1-ylbenzene, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

P22303-25G P22303-5G
SIALP22303-25GEA 167 EUR
SIALP22303-25G SIALP22303-5G
Cyclohexen-1-ylbenzene, Sigma-Aldrich®
Cyclohexen-1-ylbenzene
Formula: C₁₂H₁₄
Boiling Pt: 251 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1.004 g/cm³ (0 °C)
MDL Number: MFCD00001542
CAS Number: 771-98-2
EINECS: 212-242-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce31989847b7f5823ac4c86db68428fc148afb1d&vid=eeeb1ffa4c17dd4e58f5fc1f819153e435054e02&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR