4-Phenylcyclohexanone, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Phenylcyclohexan-1-one, 1-Oxo-4-phenylcyclohexane
M-Clarity™ quality level = MQ200
196231-10G 196231-50G
SIAL196231-10GEA 55.1 EUR
SIAL196231-10G SIAL196231-50G
4-Phenylcyclohexanone, Sigma-Aldrich®
4-Phenylcyclohexanone
Formula: C₁₂H₁₄O
MW: 174.24 g/mol
Boiling Pt: 294
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00001641
CAS Number: 4894-75-1
EINECS: 225-517-0