o-Terphenyl, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
T2800-25G T2800-100G
SIALT2800-25GEA 109 EUR
SIALT2800-25G SIALT2800-100G
o-Terphenyl, Sigma-Aldrich®
o-Terphenyl
Formula: C₁₈H₁₄
Boiling Pt: 338 °C (1013 hPa)
Melting Pt: 54…56 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 163 °C
Storage Temperature: Ambient
MDL Number: MFCD00003055
CAS Number: 84-15-1
EINECS: 201-517-6