7-Amino-4-methylcoumarin 98%
Supplier: Thermo Fisher Scientific
Warning
218060010 218060050 218062500
ACRO218060010EA 142 EUR
ACRO218060010 ACRO218060050 ACRO218062500
7-Amino-4-methylcoumarin 98%
7-Amino-4-methylcoumarin
Formula: C₁₀H₉NO₂
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 223…226 °C
Density: 1,377 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006868
CAS Number: 26093-31-2
EINECS: 247-454-8
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Yellow to beige to brown Powder
Infrared spectrum Conforms
HPLC ≥97.5 %