(1S)-(-)-α-Pinene, Sigma-Aldrich®

Supplier: Merck

Synonyms: (-)-α-Pinene, (1S)-(-)-2-Pinene, (-)-Pin-2(3)-ene, L(-)-α-Pinene

M-Clarity™ quality level = MQ300

W290203-1KG-K W290203-20KG-K W290203-8KG-K
SIALW290203-1KG-KEA 73.9 EUR
SIALW290203-1KG-K SIALW290203-20KG-K SIALW290203-8KG-K
(1S)-(-)-α-Pinene, Sigma-Aldrich®
(1S)-(-)-α-Pinene
Formula: C₁₀H₁₆
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: –62 °C
Density: 0.8592 g/cm³ (20 °C)
Flash Pt: 32 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064145
CAS Number: 7785-26-4
EINECS: 232-077-3
UN: 2368
ADR: 3,III
Merck Index: 12,07599

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR