1,5-Diphenylcarbazide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Diphenylcarbazide, sym-Diphenylcarbazide, 1,5-Diphenylcarbohydrazide, 1,5-Diphenylcarbonohydrazide, N'',N'''-Diphenylcarbonohydrazide, sym. Diphenylcarbazide
D7766-100G D7766-10G D7766-25G
SIALD7766-100GEA 207 EUR
SIALD7766-100G SIALD7766-10G SIALD7766-25G
1,5-Diphenylcarbazide, Sigma-Aldrich®
1,5-Diphenylcarbazide
Formula: C₁₃H₁₄N₄O
MW: 242.28 g/mol
Boiling Pt: 454
Melting Pt: 170…172 °C
Density: 1.31 g/cm³ (20 °C)
MDL Number: MFCD00003013
CAS Number: 140-22-7
EINECS: 205-403-7
Merck Index: 13,03353
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03cf8836a36f1077443ec2e3cfa151621925bd46&vid=a1b2bd876c3fbd9d6ded95c8430e4ec16bf5172b&prid=web&clid=SAPDAM