2-Phenylquinoline, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ200
299650-1G
299650-5G
SIAL299650-1GEA
74
EUR
SIAL299650-1G
SIAL299650-5G
2-Phenylquinoline, Sigma-Aldrich®
2-Phenylquinoline
Formula:
C₁₅H₁₁N Boiling Pt: 363 °C (1013 hPa) Melting Pt: 80 °C |
MDL Number:
MFCD00011568 CAS Number: 612-96-4 EINECS: 210-326-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...