1,3-Cyclohexadiene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
C100005-100ML C100005-25ML C100005-5ML
SIALC100005-100MLEA 751 EUR
SIALC100005-100ML SIALC100005-25ML SIALC100005-5ML
1,3-Cyclohexadiene, Sigma-Aldrich®
1,3-Cyclohexadiene
Formula: C₆H₈
Boiling Pt: 80 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.84 g/cm³ (20 °C)
Flash Pt: –8 °C
Storage Temperature: Ambient
MDL Number: MFCD00001532
CAS Number: 592-57-4
EINECS: 209-764-1
UN: 3295
ADR: 3,II