2-Chlorocyclopentanone, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
C32801-25G C32801-5G
SIALC32801-25GEA 352 EUR
SIALC32801-25G SIALC32801-5G
2-Chlorocyclopentanone, Sigma-Aldrich®
2-Chlorocyclopentanone
Formula: C₅H₇ClO
Boiling Pt: 72 °C (1013 hPa)
Density: 1.2061 g/cm³ (0 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001410
CAS Number: 694-28-0
EINECS: 211-769-9
UN: 2810
ADR: 6.1,III