Cyclopentanone, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ300

W391018-10KG-K W391018-1KG-K W391018-5KG-K
SIALW391018-10KG-KEA 634 EUR
SIALW391018-10KG-K SIALW391018-1KG-K SIALW391018-5KG-K
Cyclopentanone, Sigma-Aldrich®
Cyclopentanone
Formula: C₅H₈O
Boiling Pt: 130 °C (1013 hPa)
Melting Pt: –58 °C
Density: 0.941 g/cm³ (20 °C)
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00001409
CAS Number: 120-92-3
EINECS: 204-435-9
UN: 2245
ADR: 3,III
Merck Index: 13,02771

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR