1-Aminocyclopentanecarboxylic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
1-Aminocyclopentanecarboxylic Acid, Cycloleucine, 1-Aminocyclopentane-1-carboxylic acid
M-Clarity™ quality level = MQ100
A48105-10G
A48105-5G
SIALA48105-10GEA
277
EUR
SIALA48105-10G
SIALA48105-5G
1-Aminocyclopentanecarboxylic acid, Sigma-Aldrich®
1-Aminocyclopentanecarboxylic acid
Formula:
C₆H₁₁NO₂ Boiling Pt: 254 °C (1013 hPa) Melting Pt: 163 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001381 CAS Number: 52-52-8 EINECS: 200-144-6 UN: 2811 ADR: 6.1,III Merck Index: 13,02763 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...