N,N-Dimethylcyclohexylamine 99%
Supplier: Thermo Fisher Scientific
Danger
408255000 408250025
ACRO408255000EA 57.6 EUR
ACRO408255000 ACRO408250025
N,N-Dimethylcyclohexylamine 99%
N,N-Dimethylcyclohexylamine
Formula: C₈H₁₇N
Boiling Pt: 160…161 °C (1013 hPa)
Melting Pt: –60 °C
Density: 0.84 g/cm³
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00003844
CAS Number: 98-94-2
EINECS: 202-715-5
UN: 2264
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4525 to 1.4545 (20°C, 589 nm)
Water ≤0.25 %
GC ≥98.5 %
Color scale ≤50 APHA