Cyclohexyl isothiocyanate

Supplier: Thermo Scientific

TL01156.1 TL01156.50
MAYBTL01156.1EA 0 EUR
MAYBTL01156.1 MAYBTL01156.50
Cyclohexyl isothiocyanate
Cyclohexyl isothiocyanate
Formula: C₇H₁₁NS
Boiling Pt: 219 °C (1013 hPa)
Melting Pt: 70…72 °C
Density: 1.0339 g/cm³ (20 °C)
MDL Number: MFCD00003841
CAS Number: 1122-82-3
EINECS: 214-361-9
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR