Dicyclohexylamine, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
185841-100G 185841-500G 185841-5G 185841-2KG
SIAL185841-100GEA 88.4 EUR
SIAL185841-100G SIAL185841-500G SIAL185841-5G SIAL185841-2KG
Dicyclohexylamine, Sigma-Aldrich®
Dicyclohexylamine
Formula: C₁₂H₂₃N
Boiling Pt: 255.8 °C (1013 hPa)
Melting Pt: –2 °C
Density: 0.91 g/cm³ (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00011658
CAS Number: 101-83-7
EINECS: 202-980-7
UN: 2565
ADR: 8,III
Merck Index: 13,03122