Cyclopentanol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
C112208-100ML
C112208-500ML
C112208-5ML
SIALC112208-100MLEA
51.1
EUR
SIALC112208-100ML
SIALC112208-500ML
SIALC112208-5ML
Cyclopentanol, Sigma-Aldrich®
Cyclopentanol
Formula:
C₅H₁₀O Boiling Pt: 139 °C (1013 hPa) Melting Pt: –19 °C Density: 0.949 g/cm³ (20 °C) Flash Pt: 51 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001363 CAS Number: 96-41-3 EINECS: 202-504-8 UN: 2244 ADR: 3,III Merck Index: 13,02770 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...