Ethynylcyclohexane, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
129216-1G
129216-5G
SIAL129216-1GEA
72.9
EUR
SIAL129216-1G
SIAL129216-5G
Ethynylcyclohexane, Sigma-Aldrich®
Ethynylcyclohexane
Formula:
C₈H₁₂ Boiling Pt: 130…132 °C (1013 hPa) Density: 0.8431 g/cm³ (20 °C) |
MDL Number:
MFCD00001513 CAS Number: 931-48-6 EINECS: 213-236-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...