Ethynylcyclopropane 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
369130010
369130100
369130500
ACRO369130010EA
17.6
EUR
ACRO369130010
ACRO369130100
ACRO369130500
Ethynylcyclopropane 98%
Ethynylcyclopropane
Formula:
C₅H₆ Boiling Pt: 51…52 °C Density: 0.7801 g/cm³ Flash Pt: –17 °C Storage Temperature: Ambient |
MDL Number:
MFCD02181090 CAS Number: 6746-94-7 UN: 1993 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4270 to 1.4310 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...