4-tert-Butylcyclohexanone, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
B92303-100G B92303-25G
SIALB92303-100GEA 73.5 EUR
SIALB92303-100G SIALB92303-25G
4-tert-Butylcyclohexanone, Sigma-Aldrich®
4-tert-Butylcyclohexanone
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 225
Melting Pt: 47…50 °C
Density: 0.893 g/cm³ (50 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001642
CAS Number: 98-53-3
EINECS: 202-678-5