4-Methylcyclohexanone, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
173614-100ML 173614-25ML 173614-500ML
SIAL173614-100MLEA 192 EUR
SIAL173614-100ML SIAL173614-25ML SIAL173614-500ML
4-Methylcyclohexanone, Sigma-Aldrich®
4-Methylcyclohexanone
Formula: C₇H₁₂O
MW: 112.17 g/mol
Boiling Pt: 170
Melting Pt: –41 °C
Density: 0.92 g/cm³ (20 °C)
Flash Pt: 48
Storage Temperature: Ambient
MDL Number: MFCD00001643
CAS Number: 589-92-4
EINECS: 209-665-3
UN: 2297
ADR: 3 III