2-Methyl-1,3-cyclopentanedione, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Methylcyclopentane-1,3-dione

M-Clarity™ quality level = MQ100

117021-25G 117021-5G
SIAL117021-25GEA 53.3 EUR
SIAL117021-25G SIAL117021-5G
2-Methyl-1,3-cyclopentanedione, Sigma-Aldrich®
2-Methyl-1,3-cyclopentanedione
Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 213
Density: 1.33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001406
CAS Number: 765-69-5
EINECS: 212-153-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR