1-Methylcyclohexene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
129801-100G
129801-25G
129801-5G
SIAL129801-100GEA
337
EUR
SIAL129801-100G
SIAL129801-25G
SIAL129801-5G
1-Methylcyclohexene, Sigma-Aldrich®
1-Methylcyclohexene
Formula:
C₇H₁₂ Boiling Pt: 110 °C (1013 hPa) Melting Pt: –120.4 °C Density: 0.8102 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001548 CAS Number: 591-49-1 EINECS: 209-718-0 UN: 3295 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...