4,4-Dimethyl-2-cyclohexen-1-one ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
17162.06
17162.03
17162.06EA
118
EUR
17162.06
17162.03
4,4-Dimethyl-2-cyclohexen-1-one ≥97%
4,4-Dimethyl-2-cyclohexen-1-one
Purity: 97+%
CAS No.: 1073-13-8
Molecular Formula: (CH3)2C6H6(=O)
Formula Weight: 124.18
Flash Point: 64°(148°F)
Physical Form: Liquid
Density: 0.944
Sensitivity: Air Sensitive
Boiling Point: 73-74°/14mm
MDL No.: MFCD00009695
Formula:
C₈H₁₂O Boiling Pt: 73…74 °C (14 mmHg) Density: 0.944 Flash Pt: 64 °C (148 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009695 CAS Number: 1073-13-8 |