Octaphenylcyclotetrasiloxane ≥98%
Supplier: Thermo Fisher Scientific
L06130.14 L06130.22 AB172600.0100
L06130.14EA 62.2 EUR
L06130.14 L06130.22 ABCRAB172600.0100
Octaphenylcyclotetrasiloxane ≥98%
Octaphenylcyclotetrasiloxane
Purity: 98+% CAS No.: 546-56-5 Molecular Formula: C48H40O4Si4 Formula Weight: 793.19 Melting Point: 193-200° Boiling Point: 332°/1mm MDL No.: MFCD00003268
Formula: C₄₈H₄₀O₄Si₄
Boiling Pt: 332 °C (1 mmHg)
Melting Pt: 193…200 °C
Density: 1.089 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003268
CAS Number: 546-56-5
EINECS: 208-904-9