1-Methylpiperazine, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
130001-100G 130001-500G 130001-5G
SIAL130001-100GEA 40.5 EUR
SIAL130001-100G SIAL130001-500G SIAL130001-5G
1-Methylpiperazine, Sigma-Aldrich®
1-Methylpiperazine
Formula: C₅H₁₂N₂
MW: 100.16 g/mol
Boiling Pt: 138
Melting Pt: –6 °C
Density: 0.9085 g/cm³ (20 °C)
Flash Pt: 34
Storage Temperature: Ambient
MDL Number: MFCD00005966
CAS Number: 109-01-3
EINECS: 203-639-5
UN: 2734
ADR: 8 II