1,3-Dimethylbarbituric acid, (max. 6% H₂O) ≥99% (dry weight)
Supplier: Thermo Fisher Scientific
|
Danger
|
A13692.18
A13692.0B
A13692.30
A13692.18EA
53.8
EUR
A13692.18
A13692.0B
A13692.30
1,3-Dimethylbarbituric acid, (max. 6% H₂O) ≥99% (dry weight)
1,3-Dimethylbarbituric acid
Purity: 99%
CAS No.: 769-42-6
Molecular Formula: C6H8N2O3
Formula Weight: 156.14
Physical Form: Crystalline powder
Appearance: White/cream/pale yellow
Melting Point: 121-123°
Beilstein Reference: 139810
MDL No.: MFCD00006675
Formula:
C₆H₈N₂O₃ Melting Pt: 121…123 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006675 CAS Number: 769-42-6 EINECS: 212-211-7 |