4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222), Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ200
291110-1G
291110-250MG
SIAL291110-1GEA
212
EUR
SIAL291110-1G
SIAL291110-250MG
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222), Sigma-Aldrich®
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222)
Formula:
C₁₈H₃₆N₂O₆ Melting Pt: 69…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005111 CAS Number: 23978-09-8 EINECS: 245-962-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...