Fluorene, Sigma-Aldrich®

Supplier: Merck

Synonyms: 9H-Fluorene, Diphenylenmethane

M-Clarity™ quality level = MQ100

128333-100G 128333-500G 128333-5G
SIAL128333-100GEA 76 EUR
SIAL128333-100G SIAL128333-500G SIAL128333-5G
Fluorene, Sigma-Aldrich®
Fluorene
Formula: C₁₃H₁₀
MW: 166.22 g/mol
Boiling Pt: 295
Melting Pt: 113…115 °C
Density: 1.203 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001111
CAS Number: 86-73-7
EINECS: 201-695-5
UN: 3077
ADR: 9 III
Merck Index: 13,04181

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR