9-Fluorenylmethanol 99%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
(Fluoren-9-yl)methanol, 9-Fluorenmethanol, 9-Fluorenemethanol
170930010
170930100
170930050
170930500
ACRO170930010EA
26.2
EUR
ACRO170930010
ACRO170930100
ACRO170930050
ACRO170930500
9-Fluorenylmethanol 99%
9-Fluorenylmethanol
Formula:
C₁₄H₁₂O MW: 196.25 g/mol Boiling Pt: 337 Melting Pt: 104…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001139 CAS Number: 24324-17-2 EINECS: 246-167-5 |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 104 to 107 °C |
GC | ≥98.5 % |
Specific optical rotation | +293° to +303° (20 °C, 589 nm) (c=2, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...