9-Fluorenylmethanol
Supplier: Molekula
Synonyms:
(Fluoren-9-yl)methanol, 9-Fluorenmethanol, 9-Fluorenemethanol
18629123-100G
18629123-25G
MOLE18629123-100GEA
454
EUR
MOLE18629123-100G
MOLE18629123-25G
9-Fluorenylmethanol
9-Fluorenylmethanol
Formula:
C₁₄H₁₂O MW: 196.25 g/mol Boiling Pt: 337 Melting Pt: 107 |
MDL Number:
MFCD00001139 CAS Number: 24324-17-2 EINECS: 246-167-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...