Ethylene carbonate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1,3-Dioxa-2-cyclopentanone
M-Clarity™ quality level = MQ200
E26258-100G E26258-3KG E26258-500G
SIALE26258-100GEA 22.6 EUR
SIALE26258-100G SIALE26258-3KG SIALE26258-500G
Ethylene carbonate, Sigma-Aldrich®
Ethylene carbonate
Formula: C₃H₄O₃
Boiling Pt: 243 °C (1013 hPa)
Melting Pt: 36 °C
Density: 1.47 g/cm³ (20 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00005382
CAS Number: 96-49-1
EINECS: 202-510-0