2-Furaldehyde, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2-Furfurol, Furfural
185914-2.5L 185914-1L 185914-500ML
SIAL185914-2.5LEA 145 EUR
SIAL185914-2.5L SIAL185914-1L SIAL185914-500ML
2-Furaldehyde, Sigma-Aldrich®
2-Furaldehyde
Formula: C₅H₄O₂
MW: 96.09 g/mol
Boiling Pt: 162
Melting Pt: –39 °C
Density: 1.15 g/cm³ (20 °C)
Flash Pt: 60
Storage Temperature: Ambient
MDL Number: MFCD00003229
CAS Number: 98-01-1
EINECS: 202-627-7
UN: 1199
ADR: 6.1 II
Merck Index: 13,04325