2-Furaldehyde, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2-Furfurol, Furfural
MQ100
W248908-1KG-K W248908-25KG-K W248908-5KG-K
SIALW248908-1KG-KEA 123 EUR
SIALW248908-1KG-K SIALW248908-25KG-K SIALW248908-5KG-K
2-Furaldehyde, Sigma-Aldrich®
2-Furaldehyde
Formula: C₅H₄O₂
MW: 96.09 g/mol
Boiling Pt: 162
Melting Pt: –39 °C
Density: 1.15 g/cm³ (20 °C)
Flash Pt: 60
Storage Temperature: Ambient
MDL Number: MFCD00003229
CAS Number: 98-01-1
EINECS: 202-627-7
UN: 1199
ADR: 6.1 II
Merck Index: 13,04325