5-Methyl-2-furaldehyde, Sigma-Aldrich®

Supplier: Merck

Synonyms: 5-Methylfurfural, 5-Methyl-2-Furaldehyde

M-Clarity™ quality level = MQ100

137316-100G 137316-25G 137316-5G
SIAL137316-100GEA 124 EUR
SIAL137316-100G SIAL137316-25G SIAL137316-5G
5-Methyl-2-furaldehyde, Sigma-Aldrich®
5-Methyl-2-furaldehyde
Formula: C₆H₆O₂
MW: 110.11 g/mol
Boiling Pt: 89
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 72
MDL Number: MFCD00003232
CAS Number: 620-02-0
EINECS: 210-622-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR