5-Methyl-2-furaldehyde, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W270202-100G-K
W270202-1KG-K
W270202-5KG-K
SIALW270202-100G-KEA
121
EUR
SIALW270202-100G-K
SIALW270202-1KG-K
SIALW270202-5KG-K
5-Methyl-2-furaldehyde, Sigma-Aldrich®
5-Methyl-2-furaldehyde
Formula:
C₆H₆O₂ Boiling Pt: 89 °C (1013 hPa) Density: 1.1 g/cm³ (20 °C) Flash Pt: 72 °C |
MDL Number:
MFCD00003232 CAS Number: 620-02-0 EINECS: 210-622-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...