2,5-Dimethylfuran, Sigma-Aldrich®
Supplier: Merck
177717-100ML 177717-25ML
SIAL177717-100MLEA 107 EUR
SIAL177717-100ML SIAL177717-25ML
2,5-Dimethylfuran, Sigma-Aldrich®
2,5-Dimethylfuran
Formula: C₆H₈O
Boiling Pt: 92 °C (1013 hPa)
Melting Pt: –62 °C
Density: 0.903 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003250
CAS Number: 625-86-5
EINECS: 210-914-3
UN: 3271
ADR: 3,II