Molybdic acid ≥85% (MoO₃ basis) ACS
Supplier: Thermo Fisher Scientific
Warning
22983.36 22983.A3
22983.36EA 222 EUR
22983.36 22983.A3
Molybdic acid ≥85% (MoO₃ basis) ACS
Molybdic acid
CAS No.: 7782-91-4 Physical Form: Crystalline powder Density: 3.1 Odour: Odourless MDL No.: MFCD00036287
Formula: H₂MoO₄
MW: 161.95 g/mol
Boiling Pt: ∼300 °C (1013 hPa)
Melting Pt: 300
Density: 3.1
Storage Temperature: Ambient
MDL Number: MFCD00036287
CAS Number: 7782-91-4
EINECS: 231-970-5
Merck Index: 13,06262