Water, BioReagent cell culture reagent, sterile-filtered, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W3500-100ML W3500-1L W3500-24X500ML W3500-500ML W3500-6X1L W3500-6X500ML
SIALW3500-100MLEA 16.5 EUR
SIALW3500-100ML SIALW3500-1L SIALW3500-24X500ML SIALW3500-500ML SIALW3500-6X1L SIALW3500-6X500ML
Water, BioReagent cell culture reagent, sterile-filtered, Sigma-Aldrich®
Water
This product is a very low endotoxin (less than 1 EU/mL), 0.2 micron filtered.
For use in the preparation of cell culture media, and cell suspension and washing solutions.
Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098