Methylphosphonic acid, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Methanephosphonic acid

M-Clarity™ quality level = MQ100

289868-1G 289868-25G 289868-5G
SIAL289868-1GEA 50 EUR
SIAL289868-1G SIAL289868-25G SIAL289868-5G
Methylphosphonic acid, Sigma-Aldrich®
Methylphosphonic acid
Formula: CH₅O₃P
Melting Pt: 107.5 °C
Density: 1.639 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002137
CAS Number: 993-13-5
EINECS: 213-607-2
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR