Orthophosphoric acid, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Phosphoric acid
M-Clarity™ quality level = MQ200
466123-100G
466123-25G
SIAL466123-100GEA
171
EUR
SIAL466123-100G
SIAL466123-25G
Orthophosphoric acid, Sigma-Aldrich®
Orthophosphoric acid
Formula:
H₃PO₄ Boiling Pt: ∼158 °C (1013 hPa) Melting Pt: ∼21 °C Density: 1.541 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011340 CAS Number: 7664-38-2 EINECS: 231-633-2 UN: 1805 ADR: 8,III REACH: 01-2119485924-24 Merck Index: 14,07344 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...