Orthotelluric acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: Telluric acid
M-Clarity™ quality level = MQ100
308463-100G 308463-25G
SIAL308463-100GEA 320 EUR
SIAL308463-100G SIAL308463-25G
Orthotelluric acid, Sigma-Aldrich®
ortho-Telluric acid
Formula: H₆O₆Te
Melting Pt: 136 °C
Density: ∼3.068 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011609
CAS Number: 7803-68-1
EINECS: 232-267-6
UN: 3284
ADR: 6.1,III
Merck Index: 12,09272