Lithium aluminum hydride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Aluminium lithium hydride, Lithium tetrahydridoaluminate
MQ200
199877-10G 199877-1KG 199877-25G
SIAL199877-10GEA 98.8 EUR
SIAL199877-10G SIAL199877-1KG SIAL199877-25G
Lithium aluminum hydride, Sigma-Aldrich®
Lithium aluminum hydride
Formula: H₄AlLi
MW: 37.95 g/mol
Melting Pt: 125
Density: 0.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011075
CAS Number: 16853-85-3
EINECS: 240-877-9
UN: 1410
ADR: 4.3 I
Merck Index: 13,00348