Sodium borohydride
Supplier: Roth Carl
Danger
Synonyms: Sodium tetrahydridoborate, Sodium tetrahydroborate
4051.1 4051.2
ROTH4051.1EA 50.9 EUR
ROTH4051.1 ROTH4051.2
Sodium borohydride
Sodium borohydride
Formula: NaBH₄
MW: 37.83 g/mol
Melting Pt: 400
Density: 1.07 g/cm³ (20 °C)
Flash Pt: 69
Storage Temperature: Ambient
MDL Number: MFCD00003518
CAS Number: 16940-66-2
EINECS: 241-004-4
UN: 1426
ADR: 4.3 I
Merck Index: 13,08664