Sodium borohydride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Sodium tetrahydridoborate, Sodium tetrahydroborate
452882-2KG 452882-100G 452882-25G 452882-500G 452882-6X100G 452882-6X25G
SIAL452882-2KGEA 782 EUR
SIAL452882-2KG SIAL452882-100G SIAL452882-25G SIAL452882-500G SIAL452882-6X100G SIAL452882-6X25G
Sodium borohydride, Sigma-Aldrich®
Sodium borohydride
Formula: NaBH₄
MW: 37.83 g/mol
Melting Pt: 400
Density: 1.07 g/cm³ (20 °C)
Flash Pt: 69
Storage Temperature: Ambient
MDL Number: MFCD00003518
CAS Number: 16940-66-2
EINECS: 241-004-4
UN: 1426
ADR: 4.3 I
Merck Index: 13,08664