Sodium borohydride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Sodium tetrahydridoborate, Sodium tetrahydroborate
M-Clarity™ quality level = MQ200
480886-100G 480886-25G
SIAL480886-100GEA 267 EUR
SIAL480886-100G SIAL480886-25G
Sodium borohydride, Sigma-Aldrich®
Sodium borohydride
Formula: NaBH₄
MW: 37.83 g/mol
Melting Pt: 400
Density: 1.07 g/cm³ (20 °C)
Flash Pt: 69
Storage Temperature: Ambient
MDL Number: MFCD00003518
CAS Number: 16940-66-2
EINECS: 241-004-4
UN: 1426
ADR: 4.3 I
Merck Index: 13,08664